Learn More
Catechol Dimethylether-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Catechol Dimethylether-d6 |
| CAS | 24658-24-0 |
| Molecular Formula | C8 2H6 H4 O2 |
| Quantity | 25 mg |
| Synonym | Benzene, 1,2-di(methoxy-d3)-, Benzene, o-di(methoxy-d3)- (8CI), 1,2-Di(methoxy-d3)benzene, 1,2-Bis(trideuteriomethoxy)benzene, 1,2-Dimethoxybenzene-D6 (dimethoxy-D6), 1,2-Dimethoxybenzene-d6, 2-Methoxyanisole-d6, Catechol di(methyl-d3) ether, o-Hydroquinone di(methyl-d3) ether |
| InChI Formula | InChI=1S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6H,1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])Oc1ccccc1OC([2H])([2H])[2H] |
| IUPAC Name | 1,2-bis(trideuteriomethoxy)benzene |
| Molecular Weight (g/mol) | 144.2 |
| Formula Weight | 144.1057 |