Learn More
Catechol-d4, TRC
Product Code. 30364746
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30364746 Supplier Toronto Research Chemicals Supplier No. TRCC215176100MG
 Restricted Item
Estimated Shipment: 09-10-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Catechol ring-d4
CAS 103963-58-2
Molecular Formula C6 2H4 H2 O2
Quantity 100 mg
Synonym 1,2-Benzene-3,4,5,6-d4-diol, 1,2-Benzenediol-3,4,5,6-D4, Catechol-3,4,5,6-D4, Catechol (ring-D4), Benzene-1,2-diol (ring-D4)
InChI Formula InChI=1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c(O)c(O)c1[2H]
IUPAC Name 3,4,5,6-tetradeuteriobenzene-1,2-diol
Molecular Weight (g/mol) 114.14
Formula Weight 114.0619