missing translation for 'onlineSavingsMsg'
Learn More

(+)-Catechin, TRC

Product Code. 30010963
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010963 10 g 10g
30005109 500 mg 500mg
17943105 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010963 Supplier Toronto Research Chemicals Supplier No. TRCC21750010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-23-4
Molecular Formula C15 H14 O6
Molecular Weight (g/mol) 290.27
Synonym (+)-Catechin, 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, (2R-trans)-, Catechol (8CI), (2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol, (+)-(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (+)-(2R:3S)-5,7,3',4'-Tetrahydroxyflavan-3-ol, (+)-3',4',5,7-Tetrahydroxy-2,3-trans-flavan-3-ol, (+)-Catechin, (+)-Catechol, (+)-Cianidanol, (+)-Cyanidan-3-ol, (+)-Cyanidanol, (+)-Cyanidanol-3, (2R,3S)-(+)-Catechin, 3-Cyanidanol, (+)-, Biocatechin, Catechin, Catechin (flavan), Catechinic acid, Catechol (flavan), Catechuic acid, Catergen, Cianidanol, Cyanidanol, Cyanidol, D-(+)-Catechin, D-Catechin, D-Catechol, Dexcyanidanol, HB-MC-TS 35, NSC 2819, P 70A, Sunkatol No. 1, Sunphenon 90, Teafuran 30A, Teafuran 30E, d-Catechin, trans-(+)-3,3',4',5,7-Flavanpentol
IUPAC Name (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1
Formula Weight 290.079
Percent Purity >95%
Chemical Name or Material (+)-Catechin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.