Learn More
(+)-Catechin, TRC
Product Code. 30005109
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005109 500 mg 500mg
30010963 10 g 10g
17943105 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005109 Supplier Toronto Research Chemicals Supplier No. TRCC217500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 154-23-4
Molecular Formula C15 H14 O6
Molecular Weight (g/mol) 290.27
Synonym (+)-Catechin, 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, (2R-trans)-, Catechol (8CI), (2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol, (+)-(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (+)-(2R:3S)-5,7,3',4'-Tetrahydroxyflavan-3-ol, (+)-3',4',5,7-Tetrahydroxy-2,3-trans-flavan-3-ol, (+)-Catechin, (+)-Catechol, (+)-Cianidanol, (+)-Cyanidan-3-ol, (+)-Cyanidanol, (+)-Cyanidanol-3, (2R,3S)-(+)-Catechin, 3-Cyanidanol, (+)-, Biocatechin, Catechin, Catechin (flavan), Catechinic acid, Catechol (flavan), Catechuic acid, Catergen, Cianidanol, Cyanidanol, Cyanidol, D-(+)-Catechin, D-Catechin, D-Catechol, Dexcyanidanol, HB-MC-TS 35, NSC 2819, P 70A, Sunkatol No. 1, Sunphenon 90, Teafuran 30A, Teafuran 30E, d-Catechin, trans-(+)-3,3',4',5,7-Flavanpentol
IUPAC Name (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1
Formula Weight 290.079
Percent Purity >95%
Chemical Name or Material (+)-Catechin
Grade HPLC