missing translation for 'onlineSavingsMsg'
Learn More

Casticin, MedChemExpress

Product Code. 16443876
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443876 25 mg 25mg
16429008 10 mg 10mg
16421428 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443876 Supplier Medchem Express Supplier No. HYN0516
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Casticin is a methyoxylated flavonol isolated from Viticis Fructus, with antimitotic and anti-inflammatory effect. Casticin inhibits the activation of STAT3.

Casticin is a methyoxylated flavonol isolated from Viticis Fructus, with antimitotic and anti-inflammatory effect. Casticin inhibits the activation of STAT3.

Specifications

Chemical Name or Material Casticin
CAS 479-91-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H18O8
Quantity 25 mg
Synonym Vitexicarpin
Solubility Information DMSO : 100 mg/mL (267.14 mM; Need ultrasonic)
SMILES O=C1C(OC)=C(C2=CC=C(OC)C(O)=C2)OC3=CC(OC)=C(OC)C(O)=C13
Molecular Weight (g/mol) 374.34
Formula Weight 374.34
Percent Purity 98.4%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.