Learn More
Cytiva Cassette
Product Code. 12504487 Shop All Cytiva Products
Change view
Click to view available options
For Use With:
50 mL Fraction Collector F9-C and ÄKTA Avant Tubes
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
12504487 50 mL Fraction Collector F9-C and ÄKTA Avant Tubes Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12504487 Supplier Cytiva Supplier No. 28956402
Only null left
Add to basket
Add to basket
Holds up to six tubes
Holds up to 6 tubes. Pack of two. For use with Fraction collector F9-C and ÅKTA avant.
TRUSTED_SUSTAINABILITY
Specifications
Type Cassette
Packaging Quantity 2 Pack
For Use With 50 mL Fraction Collector F9-C and ÄKTA Avant Tubes