missing translation for 'onlineSavingsMsg'
Learn More

Cashmeran, TRC

Product Code. 30132843
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132843 100 mg 100mg
17980575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132843 Supplier Toronto Research Chemicals Supplier No. TRCC184750100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33704-61-9
Molecular Formula C14 H22 O
Molecular Weight (g/mol) 206.32
Synonym 4(5H)-Indanone, 6,7-dihydro-1,1,2,3,3-pentamethyl- (8CI), 1,2,3,5,6,7-Hexahydro-1,1,2,3,3-pentamethyl-4H-inden-4-one, (±)-Cashmeran, 1,1,2,3,3-Pentamethyl-1,2,3,5,6,7-hexahydro-4H-inden-4-one, 6,7-Dihydro-1,1,2,3,3-pentamethyl-4(5H)-indanone, Cashmeran, DPMI, Musk indanone, 4H-Inden-4-one, 1,2,3,5,6,7-hexahydro-1,1,2,3,3-pentamethyl-
IUPAC Name 1,1,2,3,3-pentamethyl-2,5,6,7-tetrahydroinden-4-one
SMILES CC1C(C)(C)C2=C(C(=O)CCC2)C1(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H22O/c1-9-13(2,3)10-7-6-8-11(15)12(10)14(9,4)5/h9H,6-8H2,1-5H3
Formula Weight 206.1671
Chemical Name or Material Cashmeran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.