missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol, Mikromol™

Product Code. 17962796
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962796 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962796 Supplier Mikromol Supplier No. MM0291.000250
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carvedilol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carvedilol
CAS 72956-09-3
Analyte or Component Names Carvedilol
Shipping Condition Room Temperature
Molecular Formula C24 H26 N2 O4
Certifications/Compliance ISO 17034
Synonym Carvedilol, 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]- (9CI), 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]propan-2-ol, Artist, BM 14190, Carvediol, Coreg, DQ 2466, Dilatrend, Dimitone, Eucardic, Kredex, Querto, SKF 105517, (+/-)-Carvedilol, SKF-105517, SB-508446, SKF-105517, USP Carvedilol System Suitability Mixture
InChI Formula InChI=1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3
SMILES COc1ccccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
Molecular Weight (g/mol) 406.47
Quantity 250 mg
Formula Weight 406.1893 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.