Learn More
Carvedilol Bisalkylpyrocatechol Impurity, TRC
Product Code. 30359070
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359070 25 mg 25mg
30349126 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30359070 Supplier Toronto Research Chemicals Supplier No. TRCC18465525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1346602-98-9
Molecular Formula C40H42N4O6
Molecular Weight (g/mol) 674.78
Synonym 3,3-{2,2-[1,2-Phenylenebis(oxy)]bis(ethane-2,1-diyl)}bis(azanediyl)bis(1-(9H-carbazol-4-yloxy)propan-2-ol)
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-[2-[2-[2-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]phenoxy]ethylamino]propan-2-ol
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC=C3OCC(CNCCOC4=CC=CC=C4OCCNCC(COC5=CC=CC6=C5C7=CC=CC=C7N6)O)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C40H42N4O6/c45-27(25-49-37-17-7-13-33-39(37)29-9-1-3-11-31(29)43-33)23-41-19-21-47-35-15-5-6-16-36(35)48-22-20-42-24-28(46)26-50-38-18-8-14-34-40(38)30-10-2-4-12-32(30)44-34/h1-18,27-28,41-46H,19-26H2
Chemical Name or Material Carvedilol Bisalkylpyrocatechol Impurity