CAS RN 1346602-98-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carvedilol Bisalkylpyrocatechol Impurity, TRC
CAS: 1346602-98-9 Molecular Formula: C40H42N4O6 Molecular Weight (g/mol): 674.78 Synonym: 3,3-{2,2-[1,2-Phenylenebis(oxy)]bis(ethane-2,1-diyl)}bis(azanediyl)bis(1-(9H-carbazol-4-yloxy)propan-2-ol) IUPAC Name: 1-(9H-carbazol-4-yloxy)-3-[2-[2-[2-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]phenoxy]ethylamino]propan-2-ol SMILES: C1=CC=C2C(=C1)C3=C(N2)C=CC=C3OCC(CNCCOC4=CC=CC=C4OCCNCC(COC5=CC=CC6=C5C7=CC=CC=C7N6)O)O