missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carvedilol

Product Code. 15832464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15832464 1 g 1g
15842464 5 g 5g
15852464 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15832464 Supplier Thermo Scientific Chemicals Supplier No. 462450010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72956-09-3 | C24H26N2O4 | 406.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72956-09-3
Molecular Formula C24H26N2O4
Molecular Weight (g/mol) 406.48
MDL Number MFCD00864692,MFCD00869663
InChI Key OGHNVEJMJSYVRP-UHFFFAOYNA-N
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
SMILES COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1

Specifications

Melting Point 114.0°C to 115.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very soluble in dimethyl sulfoxide, soluble in methanol & methylene chloride, slightly soluble in ether, ethanol & isopropanol
Formula Weight 406.47
Chemical Name or Material Carvedilol
compliance-icons
Product Identifier
  • Carvedilol
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Carvedilol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.