missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30457998
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457998 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457998 Supplier LoGiCal Supplier No. LGCAMP0291.0001
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Carvedilol 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carvedilol
CAS 72956-09-3
Analyte or Component Names Carvedilol
Shipping Condition Room Temperature
Molecular Formula C24 H26 N2 O4
Certifications/Compliance ISO 17034
Synonym Carvedilol, 2-Propanol, 1-(9 H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]- (9 CI), 1-(9 H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]propan-2-ol, Artist, BM 14190, Carvediol, Coreg, DQ 2466, Dilatrend, Dimitone, Eucardic, Kredex, Querto, SKF 105517, (+/-)-Carvedilol, SKF-105517, SB-508446, SKF-105517, USP Carvedilol System Suitability Mixture
InChI Formula InChI=1 S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27 H,13-16H2,1H3
SMILES COc1ccccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23
IUPAC Name 1-(9 H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 406.47
Quantity 1 mL
Formula Weight 406.1893 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.