missing translation for 'onlineSavingsMsg'
Learn More

Carpropamide, TRC

Product Code. 30134250
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134250 10 mg 10mg
17984165 25 mg 25mg
17932845 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134250 Supplier Toronto Research Chemicals Supplier No. TRCC18436310MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104030-54-8
Molecular Formula C15 H18 Cl3 N O
Molecular Weight (g/mol) 334.67
Synonym 2,2-Dichloro-N-[1-(4-chlorophenyl)ethyl]-1-ethyl-3-methylcyclopropanecarboxamide, Carpropamid, KTU 3616, Protiga, WIN
IUPAC Name 2,2-dichloro-N-[1-(4-chlorophenyl)ethyl]-1-ethyl-3-methylcyclopropane-1-carboxamide
SMILES CCC1(C(C)C1(Cl)Cl)C(=O)NC(C)c2ccc(Cl)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H18Cl3NO/c1-4-14(10(3)15(14,17)18)13(20)19-9(2)11-5-7-12(16)8-6-11/h5-10H,4H2,1-3H3,(H,19,20)
Formula Weight 333.0454
Chemical Name or Material Carpropamid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.