Learn More
Fisherbrand™ Universal Carousel Pipette Stand
Product Code. 15650785 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15650785 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15650785
  • ONLINE EXCLUSIVE PRICE
    88.60 EUR
    84.00 EUR / Pack of 1
    Save 4.60 EUR
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Holds up to 8 single channel, 4 multi channel, or a combination of both pipettes. Fisherbrand™ Universal Carousel Pipette Stand will securely hold virtually every known brand of pipette in the marketplace making it truly universal.
  • Ideal for labs that do not wish to be tied into a specific brand of pipette, or already have multiple brands in use
  • Weighted base allows for easy removal or replacement of pipettes, with one hand
  • Pipettes are held in a vertical position to help prevent contamination
  • Conveniently rotate stand head to select or store pipettes
  • Quick grip handle to move pipette stand (loaded or unloaded)
  • The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
  • Made of ABS (Acrylonitrile Butadiene Styrene)
Specifications
Autoclavable Not Autoclavable
Holds Single-channel Pipettes (8) or Multi-channel Pipettes (4)
Material ABS
Positions 8
Shape Circular
Stackable No
Sterility Non-sterile
Quantity 1 Each
Product Type Pipette Stand
Height (Metric) 408 mm