missing translation for 'onlineSavingsMsg'
Learn More

Carnitine Orotate, TRC

Product Code. 30362952
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362952 1 g 1g
30359495 10 g -
30360904 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362952 Supplier Toronto Research Chemicals Supplier No. TRCC1841651G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32543-38-7
Molecular Formula C7 H16 N O3 . C5 H3 N2 O4
Molecular Weight (g/mol) 317.3
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, 1,2,3,6-tetrahydro-2,6-dioxo-4-pyrimidinecarboxylate (1:1), (3-Carboxy-2-hydroxypropyl)trimethylammonium orotate (7CI), 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, salt with 1,2,3,6-tetrahydro-2,6-dioxo-4-pyrimidinecarboxylic acid (1:1) (9CI), Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, orotate (8CI), Carnitine orotate, DL-Carnitine Orotate
IUPAC Name (3-carboxy-2-hydroxypropyl)-trimethylazanium;2,4-dioxo-1H-pyrimidine-6-carboxylate
SMILES C[N+](C)(C)CC(O)CC(=O)O.[O-]C(=O)C1=CC(=O)NC(=O)N1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H15NO3.C5H4N2O4/c1-8(2,3)5-6(9)4-7(10)11;8-3-1-2(4(9)10)6-5(11)7-3/h6,9H,4-5H2,1-3H3;1H,(H,9,10)(H2,6,7,8,11)
Formula Weight 317.1223
Chemical Name or Material DL-Carnitine Orotate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.