missing translation for 'onlineSavingsMsg'
Learn More

Carnidazole, TRC

Product Code. 17963315
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963315 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17963315 Supplier Toronto Research Chemicals Supplier No. TRCC2422201MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42116-76-7
Molecular Formula C8 H12 N4 O3 S
Molecular Weight (g/mol) 244.27
Synonym N-[2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethyl]carbamothioic Acid O-Methyl Ester, Carbamothioic acid, [2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethyl]-, O-methyl ester (9CI), Carnidazole, Me 108, NSC 293873, R 25831, Spartrix
IUPAC Name O-methyl N-[2-(2-methyl-5-nitroimidazol-1-yl)ethyl]carbamothioate
SMILES COC(=S)NCCn1c(C)ncc1[N+](=O)[O-]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C8H12N4O3S/c1-6-10-5-7(12(13)14)11(6)4-3-9-8(16)15-2/h5H,3-4H2,1-2H3,(H,9,16)
Formula Weight 244.063
Chemical Name or Material Carnidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.