missing translation for 'onlineSavingsMsg'
Learn More

Carmustine, MedChemExpress

Product Code. 16491917
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491917 50 mg 50mg
16445768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491917 Supplier Medchem Express Supplier No. HY13585
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Carmustine is an antitumor chemotherapeutic agent, which works by akylating DNA and RNA.

Carmustine is an antitumor chemotherapeutic agent, which works by akylating DNA and RNA.

Specifications

Chemical Name or Material Carmustine
Physical Form Solid
CAS 154-93-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H9Cl2N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 100 mg/mL (467.18 mM; Need ultrasonic) ∣DMSO : ≥ 35 mg/mL (163.51 mM)
SMILES O=C(NCCCl)N(CCCl)N=O
Molecular Weight (g/mol) 214.05
Formula Weight 214.05
Percent Purity 99.29%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.