missing translation for 'onlineSavingsMsg'
Learn More

CARM1-IN-1 hydrochloride, MedChemExpress

Product Code. 16488037
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488037 5 mg 5mg
16471527 50 mg 50mg
16435328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488037 Supplier Medchem Express Supplier No. HY12759A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CARM1-IN-1 hydrochloride is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM; shows very low activity against PRMT1 and SET7(IC50 > 600 uM).

CARM1-IN-1 hydrochloride is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM

Specifications

Chemical Name or Material CARM1-IN-1 hydrochloride
CAS 2070018-31-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H22Br2ClNO3
Quantity 5 mg
Solubility Information DMSO : ≥ 38 mg/mL (64.22 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [H]Cl.O=C1/C(CN(CC2=CC=CC=C2)C/C1=C\C3=CC=C(O)C(Br)=C3)=C/C4=CC=C(O)C(Br)=C4
Molecular Weight (g/mol) 591.72
Formula Weight 591.72
Percent Purity 95.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.