missing translation for 'onlineSavingsMsg'
Learn More

Careez Solution(I), Chem Lab™

Product Code. 11985528
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11985528 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985528 Supplier Chem-Lab Supplier No. CL02.0302.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
Composition 219.5 g (CH3COO)2Zn.2H2O / l H2O
Solubility Information Water Soluble
Odor Odorless
pH ±4
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Carrez (I) solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: carrez(I)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.