Learn More
Wesbart™ Cardboard Cryoboxes
Product Code. 17165034
Change view
Click to view available options
Holds:
100 Cryovials (1 to 2 mL)
81 Cryovials (1 to 2 mL)
Dimensions (L x W x H):
133 x 133 x 35 mm
133 x 133 x 50 mm
Tube Array:
10 x 10
9 x 9
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) Tube Array Unit Size
17165034 81 Cryovials (1 to 2 mL) 133 x 133 x 35 mm 9 x 9 Each
17155034 100 Cryovials (1 to 2 mL) 133 x 133 x 50 mm 10 x 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17165034 Supplier Wesbart™ Supplier No. A1/81
  • ONLINE EXCLUSIVE PRICE
    3.81 EUR
    2.53 EUR / Pack of 1
    Save 1.28 EUR
    33% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-12-2026
Wesbart cryogenic boxes are made from fibreboard cardboard and are water-repellent for longer durability.
  • Cardboard boxes are specially made for the aluminum freezer racks.
  • Dividers are available in different diameters.
Specifications
Description Cardboard Box with Dividers
Color White
Material Cardboard
Shape Rectangular
Type Cryo Box
Holds 81 Cryovials (1 to 2 mL)
Includes Grid Divider
Sterility Non-sterile
Tube Array 9 x 9
Dimensions (L x W x H) 133 x 133 x 35 mm
Height (Metric) 35 mm