Learn More
Ratiolab™ Cardboard Cryo Boxes with 9 x 9 Grid
Product Code. 15649788 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
81 Tubes
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15649788 81 Tubes Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15649788
  • ONLINE EXCLUSIVE PRICE
    56.00 EUR
    38.80 EUR / Pack of 10
    Save 17.20 EUR
    30% Off
 Restricted Item
Estimated Shipment: 30-11-2026
These Cardboard Cryo Boxes with 9 x 9 Grid feature a grid insert to hold cryogenic samples.
  • Constructed of cardboard
  • 9 x 9 grid insert ot store samples
Specifications
Material Cardboard
Type Cryo Box
Holds 81 Tubes
Includes 9 x 9 Grid Insert
Quantity 10 Boxes
Dimensions (L x W x H) 133 x 133 x 50 mm