Learn More
Swann-Morton™ Carbon Sterile Steel Scalpel Blade
Product Code. 11748363
Change view
Click to view available options
Size:
# 12
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11748363 # 12 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11748363 Supplier Swann-Morton™ Supplier No. 0204
  • ONLINE EXCLUSIVE PRICE
    19.45 EUR
    17.46 EUR / Pack of 100
    Save 1.99 EUR
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-01-2027
Order Info
Size
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
For Use With (Application) Suture Cutter but also for Arteriotomies, Parotid Surgeries
Size # 12
For Use With (Equipment) Fits # 3 Graduated, # 3L, # 5B, # 7, # 9, # B3 and # B3L Handles
Sterility Sterile
Quantity 10/Pk.