Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blades
Product Code. 11708373
Change view
Click to view available options
Size:
# 10
# 11
# 15
# 18
# 21
# 22
# 23
# 24
Unit Size:
Pack of 100
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11708373 # 24 Pack of 100
11728363 # 10 Pack of 100
11877550 # 18 Pack of 100
11778363 # 21 Pack of 100
11738363 # 11 Pack of 100
11758363 # 15 Pack of 100
11798363 # 23 Pack of 100
11792724 # 22 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11708373 Supplier Swann-Morton™ Supplier No. 0211
  • ONLINE EXCLUSIVE PRICE
    22.50 EUR
    20.56 EUR / Pack of 100
    Save 1.94 EUR
    8% Off
 Restricted Item
In Stock
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
  • Carbon steel to BS EN 27740
  • Sterile
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
For Use With (Application) Used for Making Long Incisions in General Surgery and also in Autopsy Procedures
Size # 24
For Use With (Equipment) The # 24 fits Handles # 4, # 4L, # 4 Graduated and # 6B
Sterility Sterile
Certifications/Compliance CE compliant 93/42/EC
Quantity 10/Pk.