Learn More
Swann-Morton™ Carbon Steel Scalpel Blade Spare
Product Code. 11320905
Change view
Click to view available options
Size:
# 1
# 2
# 3
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11320905 # 1 Pack of 50
11330905 # 2 Pack of 50
11340905 # 3 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11320905 Supplier Swann-Morton™ Supplier No. 1241
  • ONLINE EXCLUSIVE PRICE
    8.20 EUR
    7.37 EUR / Pack of 50
    Save 0.83 EUR
    10% Off
Estimated Shipment: 22-12-2026
Replacement blades for knife set 12942545
Slotted plastic handle with clamping screw to grip blade. With blades Nos. 1 and 2.
Specifications
Blade Material Carbon Steel
Size # 1
Length (Metric) 55 mm
For Use With (Equipment) SCA-500-K
Description Straight
Thickness (Metric) 0.4 mm
Quantity 50/Pk.