Learn More
Swann-Morton™ Carbon Steel Individual Foil Packs, Sterile Scalpel Blades, No. 15C
Longer, more extended cutting edge than the traditional No. 15 blade.
Brand: Swann-Morton™ 0221
Additional Details : Weight : 0.80000kg
Description
- Provides additional reach, especially when combined with the ergonomically designed B3 handle.
- Longer, more extended cutting edge than the traditional No. 15 blade.
- Used by dentists carrying out periodontal procedures.
- Used by podiatrists for nucleating corns around the webbing of the toes.
- Fitment No. 3.
- Fits handles 3, 3L, 3 graduated, 5B, 7, 9, and B3.
Specifications
Carbon Steel | |
Sterile | |
100/Box | |
Individually Packaged |