missing translation for 'onlineSavingsMsg'
Learn More

Carbinoxamine Maleate Salt, TRC

Product Code. 30136625
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
Unit Size:
100mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136625 25 g 25g
30136242 5 g 5g
18021820 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136625 Supplier Toronto Research Chemicals Supplier No. TRCC17592025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3505-38-2
Molecular Formula C16 H19 Cl N2 O . C4 H4 O4
Molecular Weight (g/mol) 406.86
Synonym Ethanamine, 2-[(4-chlorophenyl)-2-pyridinylmethoxy]-N,N-dimethyl-, (2Z)-2-butenedioate (1:1), Ethanamine, 2-[(4-chlorophenyl)-2-pyridinylmethoxy]-N,N-dimethyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[p-chloro-α-[2-(dimethylamino)ethoxy]benzyl]-, maleate (1:1) (8CI), 2-[p-Chloro-α-[2-(dimethylamino)ethoxy]benzyl]pyridine Bimaleate, 2-[p-Chloro-α-[2-(dimethylamino)ethoxy]benzyl]pyridine maleate, Allergefon, Allergefon maleate, Carbinoxamine Maleate, Ciberon, Clistin, Clistine Maleate, Cliston, Hislosine, Lergefin, NSC 62362, Paracarbinoxamine maleate, Polistin T-Caps, Rondec, Rondec DM
IUPAC Name (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H19ClN2O.C4H4O4/c1-19(2)11-12-20-16(15-5-3-4-10-18-15)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-10,16H,11-12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 406.1295
Percent Purity >95%
Chemical Name or Material Carbinoxamine Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.