missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbinoxamine maleate salt, MedChemExpress
Description
Carbinoxamine maleate salt is a histamine H1 receptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Carbinoxamine maleate salt |
| CAS | 3505-38-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23ClN2O5 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (245.78 mM; Need ultrasonic) ∣H2O : 50 mg/mL (122.89 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C(C(C2=NC=CC=C2)OCCN(C)C)C=C1.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 406.86 |
| Formula Weight | 406.86 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?