missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbinoxamine Maleate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Carbinoxamine Maleate |
| CAS | 3505-38-2 |
| Analyte or Component Names | Carbinoxamine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 Cl N2 O . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanamine, 2-[(4-chlorophenyl)-2-pyridinylmethoxy]-N,N-dimethyl-, (2Z)-2-butenedioate (1:1), Ethanamine, 2-[(4-chlorophenyl)-2-pyridinylmethoxy]-N,N-dimethyl-, (Z)-2-butenedioate (1:1), Allergefon, Allergefon maleate, Carbinoxamine Maleate, Ciberon, Clistin, Clistine Maleate, Cliston, Hislosine, Lergefin, NSC 62362, Paracarbinoxamine maleate, Polistin T-Caps, Rondec, Rondec DM, Pyridine, 2-[p-chloro-alpha-[2-(dimethylamino)ethoxy]benzyl]-, maleate (1:1) (8CI), 2-[p-Chloro-alpha-[2-(dimethylamino)ethoxy]benzyl]pyridine Bimaleate, 2-[p-Chloro-alpha-[2-(dimethylamino)ethoxy]benzyl]pyridine maleate |
| InChI Formula | InChI=1S/C16H19ClN2O.C4H4O4/c1-19(2)11-12-20-16(15-5-3-4-10-18-15)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-10,16H,11-12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Solution Type | Neat |
| SMILES | CN(C)CCOC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?