missing translation for 'onlineSavingsMsg'
Learn More

Carabron, TRC

Product Code. 30359943
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359943 5 mg 5mg
30361767 2.5 mg 2.50mg
30359210 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359943 Supplier Toronto Research Chemicals Supplier No. TRCC1775555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1748-81-8
Molecular Formula C15 H20 O3
Molecular Weight (g/mol) 248.317
Synonym (3aR,4aS,5S,5aR,6aR)-Octahydro-5a-methyl-3-methylene-5-(3-oxobutyl)-2H-cyclopropa[f]benzofuran-2-one, Carabrone, Caropesia Lactone
IUPAC Name (3aR,4aS,5S,5aR,6aR)-5a-methyl-3-methylidene-5-(3-oxobutyl)-3a,4,4a,5,6,6a-hexahydrocyclopropa[f][1]benzofuran-2-one
SMILES CC(=O)CC[C@H]1[C@@H]2C[C@H]3[C@@H](C[C@]12C)OC(=O)C3=C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H20O3/c1-8(16)4-5-11-12-6-10-9(2)14(17)18-13(10)7-15(11,12)3/h10-13H,2,4-7H2,1,3H3/t10-,11+,12+,13-,15-/m1/s1
Formula Weight 248.141
Chemical Name or Material Carabron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.