missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Factor H Affinity Matrix

Product Code. 30275226 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 30275226

Brand: Thermo Scientific™ 2944062005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CaptureSelect Factor H Affinity Matrix is designed for the purification of human complement Factor H from plasma and recombinant sources.

CaptureSelect Factor H Affinity Matrix is designed for the purification of human complement Factor H from plasma and recombinant sources.

Features of the CaptureSelect Factor H Affinity Matrix include:

  • Highly selective for human Factor H with no affinity for other human plasma proteins
  • Binds both plasma-derived and recombinant full-length Factor H
  • Enables efficient purification of Factor H with high yields and retained activity
  • Efficient elution at acidic pH
  • Excellent scalability
  • Non-animal-derived

The high selectivity and yields obtained using CaptureSelect Factor H Affinity Matrix enable a robust, fast, and efficient purification process with excellent purity in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created using a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

Matrix: agarose-based

Average particle size: 65 ± 10 μm

Ligand: CaptureSelect Factor H affinity ligand

Ligand coupling method: epoxide

Binding capacity: > 2 mg Factor H/mL resin

Binding conditions: PBS or TBS, pH 7

Elution conditions: 20 mM citric acid, pH 3

Flow characteristics: 50–200 cm/h (up to 2 bar)

Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity > 2 mg Factor H/mL resin
Flow Rate 50 to 200 cm/h (up to 2 bar)
Formulation 20% (v/v) ethanol
Content And Storage Store at 2°C to 8°C. Protect from direct daylight.
Particle Size 63 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Resin
Ligand Type CaptureSelect Factor H Affinity Ligand
Format Agarose bead
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.