Description
- High efficiency particulate filtration
- Simple, intuitive communication by light shows the status of the hood
- Internal UV light
- Air flow monitoring
- Internal LED lighting
- UV resistant front and side panels
- Ergonomic design
- Easy to clean
- Mobile or fixed benches
- Filter HP - HEPA H14 : This filtration technology traps particles larger than 0.1 μm with 99.995
- Adding a carbon filter to your enclosure allows protection of your samples from VOCs
- Filter AS for organic vapors
Specifications
Specifications
| Product Type | Mobile Ductless Fiiltering PCR Workstation |
| Air Velocity | 240 m3/h |
| Certifications/Compliance | NF EN 61010 - CE Marking - EN 1822:1998 (HEPA H14 & ULPA U16 Filters) |
| Dimensions (L x W x H) | 630 x 1030 x 945 mm |
| Includes | HEPA and AS Filter |
| Voltage | 80/230 V |
| Wattage | 75 W |
| Frequency | 50/60 Hz |