Learn More
Globe Scientific Caps for Automation
Product Code. 31922052
Change view
Click to view available options
For Use With (Equipment):
Beckman DXA5000
Roche E801
Unit Size:
Pack of 10000
Pack of 5000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
31922052 Roche E801 Pack of 5000
31922936 Beckman DXA5000 Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31922052 Supplier Globe Scientific Supplier No. 5537
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Seamless cost-effective replacements for OEM caps.
  • Globe Scientific caps for automated systems fit several of the most popular platforms and fit seamlessly into capping processes.
  • These cost-effective alternatives to OEM caps are manufactured at high standards to ensure efficient sample processing.
  • Seamless cost-effective replacements for OEM caps
  • Smooth operation in existing platforms
  • Reliable sealing during high-speed transport and agitation
  • Made from high-grade Polyethylene (PE) material
  • Non-Sterile
Specifications
Type Automated Sampling System Cap
For Use With (Equipment) Roche E801
Note: All instruments and names of manufacturers mentioned herein are the trademarks of their respective companies and are used for reference purposes only. Globe Scientific LLC. Expressly disclaims any affiliation to them