missing translation for 'onlineSavingsMsg'
Learn More

ε-Caprolactam, Solid, MP Biomedicals™

Product Code. 11459071 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459071 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459071 Supplier MP Biomedicals Supplier No. 0215492683

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105-60-2
Density 1.014g/cm3 at 80°C
Molecular Formula C6H11NO
Quantity 250 g
Synonym epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
InChI Key JBKVHLHDHHXQEQ-UHFFFAOYSA-N
SMILES C1CCC(=O)NCC1
IUPAC Name azepan-2-one
Molecular Weight (g/mol) 113.16
PubChem CID 7768
ChEBI CHEBI:28579
Formula Weight 113.2
Physical Form Solid
Chemical Name or Material ε-Caprolactam
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.