Learn More
Hirschmann™ Dispsable Capillary Tubes
Product Code. 10049640
Change view
Click to view available options
Diameter (Metric) Inner:
1.35 mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
10049640 1.35 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10049640 Supplier Hirschmann™ Supplier No. 9221275
  • ONLINE EXCLUSIVE PRICE
    178.00 EUR
    157.00 EUR / Pack of 1000
    Save 21.00 EUR
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Excellent for working with small samples in the laboratory. Sodium-heparinized disposable blood gas analysis capillary pipettes.
  • 225 i. u. Sodium
  • e.g. for blood gas analysis
  • CE acc. IVD 98/79EC
Specifications
Product Type Capillary Tube
Capacity (Metric) 100 μL
Diameter (Metric) Inner 1.35 mm
Length (Metric) 75 mm