Learn More
Emballator Cans With Snap on Handle
Product Code. 11324425
Encompass
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
Diameter (Metric) Outer:
50 mm
100 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer unitSize
11324425 10 L 100 mm Each
11344405 10 L 50 mm Each
11334405 5 L 50 mm Each
11314425 5 L 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11324425 Supplier Emballator Supplier No. 48C1000100117
Add to basket
Add to basket
Made of HDPE. Emballator Cans With Snap on Handle is available in sizes between 2-10L.
  • Available in sizes between 2 - 10L
  • Material: HDPE
  • Approved for food contact
Specifications
Product Type Can, Snap Handle
Capacity (Metric) 10 L
Color Natural
Material HDPE
Length (Metric) 235 mm
Width (Metric) 158 mm
Height (Metric) 375.8 mm
Diameter (Metric) Outer 100 mm
Weight (Metric) 430 g