Learn More
Canrenone, TRC
Product Code. 17901195
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901195 10 mg 10mg
17964415 50 mg 50mg
17960795 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17901195 Supplier Toronto Research Chemicals Supplier No. TRCC17561010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 976-71-6
Molecular Formula C22 H28 O3
Molecular Weight (g/mol) 340.46
Synonym (2'R)-3',4'-Dihydro-5'H-spiro[androst-4,6-diene-17,2'-furan]-3,5'-dione, Spironolactone Imp. F (EP), Pregna-4,6-diene-21-carboxylic acid, 17-hydroxy-3-oxo-, γ-lactone, (17α)-, 17α-Pregna-4,6-diene-21-carboxylic acid, 17-hydroxy-3-oxo-, γ-lactone (6CI,7CI,8CI), Spiro[17H-cyclopenta[a]phenanthrene-17,2'(5'H)-furan], pregna-4,6-diene-21-carboxylic acid deriv., 11614 R.P., 17-Hydroxy-3-oxo-17α-pregna-4,6-diene-21-carboxylic acid lactone, 17-Hydroxy-3-oxo-17α-pregna-4,6-diene-21-carboxylic acid γ-lactone, 17α-(2-Carboxyethyl)-17β-hydroxyandrosta-4,6-dien-3-one lactone, 17β-Hydroxy-3-oxopregna-4,6-diene-21-carboxylic acid, 20-Spiroxa-4,6-diene-3,21-dione, 3-(17β-Hydroxy-3-oxoandrosta-4,6-dien-17α-yl)propionic acid lactone, 3-(17β-Hydroxy-3-oxoandrosta-4,6-dien-17α-yl)propionic acid γ-lactone, 3-(3-Oxo-17β-hydroxy-4,6-androstadien-17α-yl)propionic acid γ-lactone, 3'-(3-Oxo-17β-hydroxyandrosta-4,6-dien-17α-yl)-propionic acid lactone, Aldadiene, Canrenone, NSC 261713, Phanurane, SC 9376, Spirolactone SC 14266
IUPAC Name (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione
SMILES C[C@]12CCC(=O)C=C1C=C[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]45CCC(=O)O5
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C22H28O3/c1-20-9-5-15(23)13-14(20)3-4-16-17(20)6-10-21(2)18(16)7-11-22(21)12-8-19(24)25-22/h3-4,13,16-18H,5-12H2,1-2H3/t16-,17+,18+,20+,21+,22-/m1/s1
Formula Weight 340.2038
Percent Purity >95%
Chemical Name or Material Canrenone
Grade HPLC