Learn More
Canrenone, 98%, Thermo Scientific™
Inhibits aldosterone biosynthesis and blocks ouabain effects
70.80 EUR - 898.47 EUR
Chemical Identifiers
| CAS | 976-71-6 |
|---|---|
| Molecular Formula | C22H28O3 |
| Molecular Weight (g/mol) | 340.463 |
| MDL Number | MFCD00071735 |
| InChI Key | UJVLDDZCTMKXJK-WNHSNXHDSA-N |
| Synonym | canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone |
| PubChem CID | 13789 |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
| SMILES | CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15493039
|
Thermo Scientific Alfa Aesar
J60238.03 |
1g | |||||||
|
15403049
|
Thermo Scientific Alfa Aesar
J60238.06 |
5g |
274.00 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15413049
|
Thermo Scientific Alfa Aesar
J60238.14 |
25g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C22H28O3 | |
| MFCD00071735 | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
Specifications
| 976-71-6 | |
| MFCD00071735 | |
| canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
| CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C | |
| 340.463 | |
| 340.46 | |
| Canrenone |
| C22H28O3 | |
| 14,1752 | |
| UJVLDDZCTMKXJK-WNHSNXHDSA-N | |
| (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione | |
| 13789 | |
| 98% |
Safety and Handling
missing translation for 'einecsNumber' : 213-554-5
missing translation for 'tsca' : No
Recommended Storage : Keep cold