Learn More
Candesartan Cilexetil Methoxy Analogue, TRC
Product Code. 30348229
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348229 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30348229 Supplier Toronto Research Chemicals Supplier No. TRCC1755851MG
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Candesartan Cilexetil Methoxy Analogue
CAS 1026042-12-5
Molecular Formula C32 H32 N6 O6
Quantity 1 mg
InChI Formula InChI=1S/C32H32N6O6/c1-20(43-32(40)44-23-9-4-3-5-10-23)42-30(39)26-13-8-14-27-28(26)38(31(33-27)41-2)19-21-15-17-22(18-16-21)24-11-6-7-12-25(24)29-34-36-37-35-29/h6-8,11-18,20,23H,3-5,9-10,19H2,1-2H3,(H,34,35,36,37)
SMILES COc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)c5ccccc5c6nnn[nH]6
IUPAC Name 1-cyclohexyloxycarbonyloxyethyl 2-methoxy-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
Molecular Weight (g/mol) 596.63
Formula Weight 596.2383