missing translation for 'onlineSavingsMsg'
Learn More

Canada Pesticide Mixture 5 ver. 2 3000 μg/mL in Ethyl Acetate, Dr. Ehrenstorfer

Product Code. 30320132
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320132 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320132 Supplier Dr. Ehrenstorfer Supplier No. DREA50000075EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxamyl∣Spiromesifen
CAS 23135-22-0, 283594-90-1
Analyte or Component Names Oxamyl∣Spiromesifen
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Ethyl Acetate
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Oxamyl 3000 μg/mL∣Spiromesifen 3000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.