Learn More
Thermo Scientific Chemicals cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Product Code. 15450587
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450587 1 mg 1mg
15440587 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15450587 Supplier Thermo Scientific Chemicals Supplier No. J66438.M^
 Restricted Item
In Stock
Add to basket
For cellular and molecular biology applications
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24) acts as inhibitor of c-AMP-dependent protein kinase These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2222.40
Product Type cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp