missing translation for 'onlineSavingsMsg'
Learn More

Calyculin A, >98%, MP Biomedicals™

Product Code. 11419841 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419841 50 μg 50µg
11409841 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419841 Supplier MP Biomedicals Supplier No. 0215837850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101932-71-2
Molecular Formula C50H81N4O15P
Molecular Weight (g/mol) 1009.185
InChI Key FKAWLXNLHHIHLA-OROLNYDDSA-N
Synonym calyculin a
PubChem CID 131632568
IUPAC Name [(2R,3R,5R,7R,8R,9S)-2-[(1S,3S,4S,5R,6R)-14-cyano-3,5-dihydroxy-1-methoxy-4,6,8,9,13-pentamethyltetradeca-7,9,11,13-tetraenyl]-9-[3-[2-[(2S)-4-[[(2S,3S,4S)-4-(dimethylamino)-2,3-dihydroxy-5-methoxypentanoyl]amino]butan-2-yl]-1,3-oxazol-4-yl]prop-2-enyl]-7
SMILES CC1C(CC2(C(C(C(O2)C(CC(C(C)C(C(C)C=C(C)C(=CC=CC(=CC#N)C)C)O)O)OC)OP(=O)(O)O)(C)C)OC1CC=CC3=COC(=N3)C(C)CCNC(=O)C(C(C(COC)N(C)C)O)O)O

Specifications

Assay Percent Range >98%
Quantity 50 μg
Formula Weight 1009.18
Physical Form Lyophilized Powder
Chemical Name or Material Calyculin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.