Learn More
California Solvent Mixture Version 2 1000 μg/mL in Triacetin Second Source, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 107-06-2, 110-71-4, 123-91-1, 71-23-8, 75-83-2, 79-29-8 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Triacetin |