Description
- For use with TB1 Portable Turbidimeter
Specifications
Specifications
| Type | Calibration Kit |
| For Use With (Equipment) | TB1 Turbidimeter |
| Includes | 800, 100, 20.0 & 0.02 NTU standard |
| For Use With (Application) | Measures the turbidity of aqueous samples simply and accurately |