Learn More
Calcium Sulfate Dihydrate, Supelco™
Specifications
Specifications
| CAS | 10101-41-4 |
| Quantity | 500 g |
| MDL Number | MFCD00149625 |
| Synonym | Calcium sulfate dihydrate, Calcium sulfate dihydrate |
| Molecular Weight (g/mol) | 172.17 |
| Grade | Research |
| Density | 2.32 g/cm3 at 20°C |
| Chemical Name or Material | Calcium Sulfate Dihydrate |