Learn More
Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Product Code. 11460512 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460512 1 kg 1kg
11440512 100 g 100g
11450512 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11460512 Supplier MP Biomedicals Supplier No. 0219141491
  • ONLINE EXCLUSIVE PRICE
    197.00 EUR
    186.90 EUR / 1kg
    Save 10.10 EUR
    5% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Chemical Identifiers
CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O
Specifications
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 98.0-102.0%
Formula Weight 172.2
Grade ACS Reagent
Chemical Name or Material Calcium Sulfate Dihydrate