Learn More
Calcium Solution for ICP, 1g/L, HCl Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12943374 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12943374 100 mL 100mL
12953374 250 mL 250mL
12963374 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12943374 Supplier Fisher Chemical Supplier No. S 009 100
  • ONLINE EXCLUSIVE PRICE
    58.65 EUR
    47.12 EUR / 100mL
    Save 11.53 EUR
    19% Off
 Restricted Item
Estimated Shipment: 04-12-2026
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains HCl
Concentration 1 g/L
Quantity 100 mL