missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, Supelco™

Product Code. 15814200
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814200 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15814200 Supplier Supelco Supplier No. 1021205000
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

EMPLURA™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 45°C
CAS 13477-34-4
Quantity 5 kg
Molecular Formula Ca(NO3)2 · 4 H2O
MDL Number MFCD00149604
Synonym Calcium nitrate tetrahydrate, Calcium nitrate tetrahydrate
Molecular Weight (g/mol) 236.15
Grade Research
Density 1.82 g/cm3 at 20°C
Chemical Name or Material Calcium Nitrate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.