Learn More
Calcium nitrate tetrahydrate, MilliporeSigma™
Product Code. 15834200
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fibre carton
Plastic Bottle
Unit Size:
500g
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15834200 50 kg Fibre carton 50kg
15824200 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15834200 Supplier Supelco Supplier No. 1021219050
 Restricted Item
Estimated Shipment: 18-11-2026
For analysis EMSURE™ ACS
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O
Specifications
Quantity 50 kg
Odor Odorless
Packaging Fibre carton
Density 1.82g/cm3 at 20°C
UN Number UN1454
Solubility Information Soluble in water: 2710g/L at 40°C
Formula Weight 236.15
Chemical Name or Material Calcium nitrate tetrahydrate