missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, 99.95%, Suprapur™, MilliporeSigma™

Product Code. 15827080
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15827080 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15827080 Supplier Supelco Supplier No. 1021230100
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity grade salt suitable for inorganic trace analysis.

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 45°C
CAS 13477-34-4
Color Colorless
pH 5.0 to 7.0 (50g/L, H2O, 25°C)
Physical Form Solid
Quantity 100 g
Packaging Plastic Bottle
Density 1.82 g/3 (20 °C)
Molecular Formula CaH8N2O10
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
Solubility Information 2710g/L (at 40°C)
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
SMILES O.[Ca++].[O-][N+]([O-])=O
IUPAC Name calcium bis(nitric acid) tetrahydrate
Molecular Weight (g/mol) 236.15
PubChem CID 16211656
ChEBI CHEBI:86159
Percent Purity >99.995%
Grade Suprapur
Chemical Name or Material Calcium Nitrate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.