Learn More
Calcium Hypochlorite, Sigma-Aldrich™
Description
Calcium hypochlorite [Ca(OCl)2] is a stable oxidizing agent used for the conversion of ethers to esters; thioethers to sulfoxides; aldehydes to acids and alkenes to chlorohydrins. It is also used in the C-C bond cleavage and chlorination reactions.
Specifications
Specifications
| Melting Point | 100°C |
| CAS | 7778-54-3 |
| Quantity | 100 g |
| Density | 2.35 g/cm3 at 20°C |
| Molecular Formula | Ca(OCl)2 |
| MDL Number | MFCD00010900 |
| Synonym | Calcium hypochlorite, Calcium hypochlorite |
| Molecular Weight (g/mol) | 142.98 |
| Grade | Research |
| Chemical Name or Material | Calcium Hypochlorite |